missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ cis-6-Hydroxynorketamine hydrochloride

Product Code. p-200050936
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18036002

Brand: Tocris Bioscience™ 5982/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Enhances AMPA currents: antidepressant

cis-6-Hydroxynorketamine hydrochloride is a metabolite of ketamine . Increases AMPA receptor-mediated excitatory post-synaptic potentials in the CA1 region of hippocampal slices. Exhibits antidepressive effects in vivo. 2S,6S-enantiomer and 2R,6R-enantiomer also available.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material cis-2-Amino-2-(2-chlorophenyl)-6-hydroxycyclohexanone hydrochloride
CAS 2319601-04-0
Quantity 50 mg
Target Additional NMDA Receptor Compounds
Molecular Formula C12H14ClNO2.HCl
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.