missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ cis-N-Boc-4-cyano-L-proline methyl ester, 97%

Brand:  Alfa Aesar™ H52783.03

Additional Details : CAS : 487048-28-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15409527

  • 965.00€ / 1g


Estimated Shipment
14-05-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

487048-28-2
254.286
GHLKJIQVORAVHE-BDAKNGLRSA-N
17750454
CC(C)(C)OC(=O)N1CC(CC1C(=O)OC)C#N
C12H18N2O4
MFCD03787928
n-boc-cis-4-cyano-l-proline methyl ester, 2s,4s-1-tert-butyl 2-methyl 4-cyanopyrrolidine-1,2-dicarboxylate, boc-cis-4-cyano-l-proline methyl ester, 1-tert-butyl 2-methyl 2s,4s-4-cyanopyrrolidine-1,2-dicarboxylate, 1,2-pyrrolidinedicarboxylic acid, 4-cyano-, 1-1,1-dimethylethyl 2-methyl ester, 2s,4s, 1,2-pyrrolidinedicarboxylicacid, 4-cyano-, 1-1,1-dimethylethyl 2-methyl ester, 2s,4s, cis-n-boc-4-cyano-l-proline methyl ester 250mg, 2s,4s-1-tert-butoxycarbonyl-4-cyano-2-methylpyrrolidine-2-carboxylic acid, 2s,4s-4-cyano-1,2-pyrrolidinedicarboxylic acid 1-tert-butyl 2-methyl ester, 1,2-pyrrolidinedicarboxylic acid,4-cyano-,1-1,1-dimethylethyl 2-methyl ester, 2s,4s
1-O-tert-butyl 2-O-methyl (2S,4S)-4-cyanopyrrolidine-1,2-dicarboxylate

Specifications

cis-N-Boc-4-cyano-L-proline methyl ester
487048-28-2
MFCD03787928
GHLKJIQVORAVHE-BDAKNGLRSA-N
1-O-tert-butyl 2-O-methyl (2S,4S)-4-cyanopyrrolidine-1,2-dicarboxylate
17750454
97%
1g
C12H18N2O4
n-boc-cis-4-cyano-l-proline methyl ester, 2s,4s-1-tert-butyl 2-methyl 4-cyanopyrrolidine-1,2-dicarboxylate, boc-cis-4-cyano-l-proline methyl ester, 1-tert-butyl 2-methyl 2s,4s-4-cyanopyrrolidine-1,2-dicarboxylate, 1,2-pyrrolidinedicarboxylic acid, 4-cyano-, 1-1,1-dimethylethyl 2-methyl ester, 2s,4s, 1,2-pyrrolidinedicarboxylicacid, 4-cyano-, 1-1,1-dimethylethyl 2-methyl ester, 2s,4s, cis-n-boc-4-cyano-l-proline methyl ester 250mg, 2s,4s-1-tert-butoxycarbonyl-4-cyano-2-methylpyrrolidine-2-carboxylic acid, 2s,4s-4-cyano-1,2-pyrrolidinedicarboxylic acid 1-tert-butyl 2-methyl ester, 1,2-pyrrolidinedicarboxylic acid,4-cyano-,1-1,1-dimethylethyl 2-methyl ester, 2s,4s
CC(C)(C)OC(=O)N1CC(CC1C(=O)OC)C#N
254.286
254.29
Videos
SDS Documents