missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Citrate, 0.5M buffer soln., pH 4.5

Product Code. 15482869
Change view
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
Product Code. Quantity unitSize
15482869 500 mL 500mL
15472869 250 mL 250mL
2 options
This item is not returnable. View return policy

Product Code. 15482869

Brand: Thermo Scientific Chemicals J60024.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6132-04-3294.10 g/mol

Citric acid is used as a substrate for citrate lyase, a buffer component, an anticoagulant. For anticoagulation use it is typically used at a concentration of approximately 0.129 M.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Citric acid is used as a substrate for citrate lyase, a buffer component, an anticoagulant. For anticoagulation use it is typically used at a concentration of approximately 0.129 M.

Solubility
Fully miscible with water. Insoluble in ethanol.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4-3-6132
Molecular Weight (g/mol) 294.10

Specifications

Color Colorless
Concentration 0.5M buffer soln.
pH 4.5
Physical Form Liquid
Quantity 500 mL
Odor Odorless

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.