Learn More
Abnova™ CKS1B (Human) Recombinant Protein
Product Code. 16133251
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16133251
Brand: Abnova™ H00001163P01.10ug
Human CKS1B full-length ORF ( AAH07751, 1 a.a. - 79 a.a.) recombinant protein with GST-tag at N-terminal.
  • Sequence: MSHKQIYYSDKYDDEEFEYRHVMLPKDIAKLVPKTHLMSESEWRNLGVQQSQGWVHYMIHEPEPHILLFRRPLPKKPKK
Specifications
Accession Number AAH07751
Gene ID (Entrez) 1163
Name CDC28 protein kinase regulatory subunit 1B
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias CKS1, PNAS-16, PNAS-18, ckshs1
Gene Symbol CKS1B
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer