Learn More
Hirschmann™ Class A Volumetric Pipettes with Amber Stain Graduation, USP
Product Code. 12851783
Click to view available options
Volume (Metric):
4 mL
6 mL
7 mL
10 mL
15 mL
Tolerance:
0.001 mL
0.01 mL
0.02 mL
0.03 mL
Length (Metric):
400 mm
419 mm
440 mm
500 mm
Unit Size:
Pack of 6
Product Code. 12851783
Brand: Hirschmann™ 134016427
Only null left
Add to basket
Add to basket
Provides an extremely high degree of volumetric accuracy according to USP standards for your pipetting needs. Hirschmann™ Class A Volumetric Pipettes with Amber Stain Graduation are manufactured from clear AR-Glas™.
  • Acc. to USP
  • One mark
  • With dated batch identification
Specifications
Class A
Material Soda Lime Glass
Graduated Yes
Graduation Color Brown
Quantity 6/Pk.
Tolerance 0.03 mL
Volume (Metric) 15 mL
Length (Metric) 500 mm
Type Measuring Pipet