Learn More
Hirschmann™ Class A Volumetric Pipettes with Amber Stain Graduation, USP
Click to view available options
Volume (Metric):
4 mL
6 mL
7 mL
10 mL
15 mL
Tolerance:
0.001 mL
0.01 mL
0.02 mL
0.03 mL
Length (Metric):
400 mm
419 mm
440 mm
500 mm
Unit Size:
Pack of 6
Description
- Acc. to USP
- One mark
- With dated batch identification
Specifications
Specifications
| Class | A |
| Material | Soda Lime Glass |
| Graduated | Yes |
| Graduation Color | Brown |
| Quantity | 6/Pk. |
| Tolerance | 0.001 mL |
| Volume (Metric) | 4 mL |
| Length (Metric) | 400 mm |
| Type | Measuring Pipet |