Learn More
Fisherbrand™ Class B Glass Tall Form Measuring Cylinders
Product Code. p-8000749
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
500 mL
1000 mL
2000 mL
Graduations:
0.1 mL
0.2 mL
0.5 mL
1 mL
5 mL
10 mL
20 mL
Unit Size:
Each
Product Code. 11847501
Brand: Fisherbrand™ DBG508
Only null left
Add to basket
Edge
Add to basket
Measure and pour liquids with precision with these cylinders. Fisherbrand™ Class B Glass Tall Form Measuring Cylinders are manufactured from calibrated, clear borosilicate glass.
  • Graduated
  • Stable hexagonal foot
  • Corrosion-free and resistant to thermal shock
Specifications
Certifications/Compliance ISO 4788
Class B
Height (Metric) 464 mm
Tolerance Metric ±10 mL
Type Graduated Measuring Cylinder
Material Borosilicate Glass
Diameter (Metric) Outside 67 mm
Capacity (Metric) 1000 mL
Graduations 10 mL
Autoclavable Autoclavable
Limit of Error ±10mL
Max. Temperature 500°C