Learn More
Hirschmann™ Class B Graduated Cylinders with Blue Graduations
Product Code. 13108039
Click to view available options
Unit Size:
Pack of 12
Pack of 3
Pack of 6
Capacity (Metric):
500 mL
1000 mL
Product Code. 13108039
Brand: Hirschmann™ 7501160
Only null left
Add to basket
Add to basket
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann™ Class B Measuring Cylinders, raised scale are manufactured from PP.
  • Acc. to DIN 12681/ISO 6706
  • PP
  • Raised scale
  • Blue graduated
  • Main point ring graduation
  • Hexagonal base
Specifications
Material Polypropylene
Certifications/Compliance DIN 12681, ISO 6706
Description Graduated cylinder, PP
Class B
Disposable Reusable
Diameter (Metric) 15 mm
Graduations 0.2 mL
Shape Round
For Use With (Application) Precise Liquid Measurement and Fluid Transfer in the Laboratory
Scale Color Blue
Base Type Hexagonal
Height (Metric) 145 mm
Type Graduated Measuring Cylinder