Learn More
Hirschmann™ Class B Graduated Polypropylene Cylinders with Raised Blue Graduations
Product Code. 13198029
Click to view available options
Unit Size:
Pack of 12
Pack of 3
Pack of 6
Capacity (Metric):
25 mL
1000 mL
Product Code. 13198029
Brand: Hirschmann™ 7500194
Only null left
Add to basket
Add to basket
Excellent for precise liquid measurement and fluid transfer in the laboratory.
  • Acc. to DIN 12681/ISO 6706
  • PP
  • Raised scale
  • Main point ring graduation
  • Hexagonal base
Specifications
Class B
Disposable Reusable
Shape Round
For Use With (Application) Precise Liquid Measurement and Fluid Transfer in the Laboratory
Scale Color Blue
Base Type Hexagonal
Height (Metric) 482 mm
Type Graduated Measuring Cylinder