Learn More
VITLAB™ Class B Polypropylene Volumetric Flask
Set volumes accurately for exact measurements in lab applications with these flasks. VITLAB™ Class B Polypropylene Volumetric Flasks feature a single calibration mark that defines the calibrated volume exactly.
Brand:  VITLAB™ 677941
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11807920
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • Highly transparent
  • With ring mark individually calibrated to ‘In’
  • Class B tolerances according to DIN EN ISO 1042
  • Thermal stress up to 60°C does not permanently exceed the tolerance limits
  • To preserve markings, cleaning at no higher than 60°C is recommended
Specifications
Specifications
Yes
Polypropylene
10 mL
90 mm
6
±0.08 mL
Screw
DIN EN ISO 1042
Transparent
Polypropylene
NS 10/19 stopper
60°C
B