Learn More
Kartell™ Class B (TPX™) PMP Low Form Beakers
Click to view available options
Capacity, Metric: 50mL
Description
Crystal clear, will withstand 170°C for short periods, 150°C for continuous work. Permanent graduations molded in for reproducibility, excellent chemical resistance. Not suitable for use with stirrer hotplates. Autoclavable.
Specifications
Color | Crystal Clear |
Diameter (Metric) Outer | 42mm |
Capacity (Metric) | 50 mL |
Material | PMP |
For Use With (Application) | Suitable for Foodstuff |
Height (Metric) | 60mm |
Quantity | 10Pack |
Type | Beaker |
Autoclavable | Autoclavable |
Graduated | Yes |