Learn More
Zwiebel™ Class E2 Box Weight
Brand:  Zwiebel™ PBI23062
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11710673
  • 148.40€ / Each
Estimated Shipment: 18-06-2024
to see stock.
Specifications
5 g
±0.05 mg
For use with Weighing Instruments of Special Accuracy Class I