Learn More
Zwiebel™ Class E2 Box Weight
Click to view available options
Mass:
2 mg
10 mg
500 mg
1 g
2 g
5 g
10 g
20 g
50 g
200 g
Tolerance:
±0.006 mg
±0.008 mg
±0.025 mg
±0.03 mg
±0.04 mg
±0.05 mg
±0.06 mg
±0.08 mg
±0.1 mg
±0.3 mg
Unit Size:
Each
Specifications
Specifications
| Mass | 1 g |
| Tolerance | ±0.03 mg |
| For Use With (Application) | For use with Weighing Instruments of Special Accuracy Class I |