Learn More
Zwiebel™ Class E2 Box Weight with COFRAC Calibration Certificate
Click to view available options
Mass:
1 g
1 to 100 g
10 g
100 g
2 g
20 g
200 g
5 g
5 kg
50 g
Unit Size:
Each
Description
- Class E2 stainless steel calibration weight
- Delivered with a calibration certificate guaranteeing traceability
- In compliance with OIML R111
- Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
| Mass | 5 g |
| Tolerance | ±0.05 mg |
| For Use With (Application) | For use with Weighing Instruments of Special Accuracy Class I |