missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zwiebel™ Class M2 Weight Set
Click to view available options
Mass:
1 to 100 g
1 to 200 g
1 to 50 g
1 to 500 g
Unit Size:
Each
Description
- Class M2 stainless steel calibration weight
- In compliance with OIML R111
- Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
| Mass | 1 to 100 g |
| For Use With (Application) | For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu