Learn More
Abeba™ Classic 31011 Shoes
Product Code. 15688287
Click to view available options
Size:
41
42
43
44
45
46
Unit Size:
1 pair
Product Code. 15688287
Brand: Abeba™ 31011/45
Only null left
Add to basket
Add to basket
Animal fats, oil and fuel resistant. Leather/microfiber construction.
  • CE EN ISO 20345:2011 SB SRC
  • Microfiber black
  • Steel toe cap
  • Slip resistance PU outsole sole
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Microfiber lining and heel patch
  • Adjustable instep area
  • Buckle fastening
Specifications
Certifications/Compliance CE EN ISO 20345:2011 SB SRC
Color Black
Liner Material Microfiber
Material Microfiber
Size 45
Sole Features Slip-resistant
Sole Material Polyurethane
Style 31011
Toe Material Steel
Resistant To Animal fats, oil and fuel