Learn More
Abeba™ Classic 31011 Shoes
Animal fats, oil and fuel resistant. Leather/microfiber construction.
Brand:  Abeba™ 31011/46
Additional Details : Weight : 0.88600kg
 View more versions of this product
Product Code. 15698287
  • 90.83€ / 1 pair
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20345:2011 SB SRC
  • Microfiber black
  • Steel toe cap
  • Slip resistance PU outsole sole
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Microfiber lining and heel patch
  • Adjustable instep area
  • Buckle fastening
Specifications
Specifications
CE EN ISO 20345:2011 SB SRC
Microfiber
46
Polyurethane
Steel
Black
Microfiber
Slip-resistant
31011
Animal fats, oil and fuel