Learn More
Tenak™ Classic Half Rack for Cryogenic Freezer, 50 mm Box Height
Product Code. 11567234 Shop All TENAK Products
Click to view available options
Holds:
Boxes
Unit Size:
Each
Product Code. 11567234
Brand: Tenak™ TE21418
Only null left
Add to basket
Add to basket
Classic Half Rack equipped with a locking rod to secure the cryoboxes.
  • With a locking rod to secure the cryoboxes
  • These racks are for half sized boxes with 5 x 5 cell divider
  • Max. footprint of 83 x 83 mm
Specifications
Holds Boxes
Material Stainless Steel
Positions 12
For Use With (Equipment) Cyrogenic Freezers
Dimensions (L x W x H) 83 x 83 x 665 mm
Height (Metric) 50 mm (Box)