Learn More
Brannan™ Classic Stainless Steel Dial Oven Thermometer
Product Code. 17310560
Click to view available options
Temperature Range (Metric):
30°C to 450°F
Unit Size:
Each
Product Code. 17310560
Brand: Brannan™ 23/400/3
Only null left
Add to basket
Add to basket
Brannan Classic Stainless Steel Dial Oven Thermometer features a black temperature scale and red pointer for clear reading
  • Suitable for hanging or sitting on oven shelf
  • Using the thermometer guides accurately setting the oven control
  • Stainless Steel 430 case and pointer, aluminum dial, glass lens, bimetal sensor
Specifications
Display Dimensions 4.2 x 4.2 cm
Emissivity Yes
For Use With (Application) Measure Temperature of Oven in Use
Height (English) 2.83 in.
Height (Metric) 7.2 cm
Length (Metric) 6.3 cm
Material Stainless Steel, Aluminum
Model 23/400/3
Weight (Metric) 50.5 g
Width (English) 1.61 in.
Length (English) 2.4 in.
Temperature Range (English) 0°C to 230°C
Temperature Range (Metric) 30°C to 450°F
Calibrated No
Accuracy ±10°C and ±10°F
Display Analog
Width (Metric) 4.1 cm