missing translation for 'onlineSavingsMsg'
Learn More

Clathrin, Light Chain Mouse, Clone: SPM174, enQuireBio™

Mouse Monoclonal Antibody

199.00€ - 510.00€

Specifications

Antigen Clathrin, Light Chain
Clone SPM174
Concentration 200μg/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15979817
View Documents Promotion Details
EnquireBio
1211-MSM1X-P1
100 μg
510.00€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 05-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15969817
View Documents Promotion Details
EnquireBio
1211-MSM1X-P0
20 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

Clathrin, Light Chain
200μg/mL
Monoclonal
Mouse
10mM PBS with 0.05% BSA and 0.05% Azide
1212
Purified recombinant N-terminal fragment of human Clathrin Light Chain
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
31-44kDa
SPM174
Immunohistochemistry
Unconjugated
RUO
XP_016864515
CLTA and CLTB
IgG2b κ
Protein A/G Purified
Cytoplasmic
HeLa cells. Placenta or Prostate Carcinoma.
Videos
SDS
Documents

Documents

Product Certifications