Learn More
Smeg™ Cleaning Powder
Clean a wide range of glassware with this alkaline detergent. Smeg™ Cleaning Powder achieves excellent washing results and optimize the thermal disinfection of laboratory instruments.
Brand: Smeg™ DETERGL
Additional Details : CAS : 1310-73-2 Weight : 10.50000kg Transport : UN number : 1759 Chem class : 8 Pack group : II
Description
- Universal alkaline powder detergent with phosphates without oxidizing surfactants and without chlorine compounds
- Application field: general chemistry, food analysis, organoleptic protein
- Available in 10kg container
Specifications
Alkaline Detergent | |
Bucket | |
10 kg |
Powder | |
1 Ea. |
Safety and Handling
- smeg DETERGLASS
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- Specific target organ toxicity Category 3
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- H335-May cause respiratory irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P501-Dispose of contents/container to only when containeris empty and closed.For disposal of product residues, referto Safety Data Shee.