Learn More
Cole-Parmer Cleanroom Hose Barb Fittings, Straight Union, Natural Nylon
Product Code. 30053225 Shop All Cole-Parmer Products
Click to view available options
Hose Barb:
1/16 in. Hose Barb
3/8 in. Hose Barb
Unit Size:
Pack of 10
Product Code. 30053225
Brand: Cole-Parmer 5010817
Only null left
Add to basket
Add to basket
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Quick and easy assembly without tools. Single barb design tightly grips the tubing. RoHS compliant
Specifications
Certifications/Compliance FDA;Non-DEHP;Non-Phthalate;RoHS
Min. Operating Temperature (Metric) -23°C
Packaging Quantity 10/Pk.
Hose Barb 1/16 in. Hose Barb
Outlet Port Hose Barb Union
Sterilization Method E-beam/gamma irradiation, EtO, or autoclave up to 121°C
Color Natural