Learn More
Cole-Parmer Cleanroom Port Disk Female Luer Fitting, Flat Plate, Engage™ 8402
Product Code. 30052994 Shop All Cole-Parmer Products
Click to view available options
:
10 Port Disk Luer Fitting - No Dimple Engage 8402
Product Code. 30052994
Brand: Cole-Parmer 5011711
Only null left
Add to basket
Add to basket
Weld to single-use bags to transfer cell culture, drug compounds, and other biological fluids
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Fle in., ible, smooth flat base for easy attachment. Ultrasonic, radio frequency, or thermal weld to polyethylene bags. Connect to a male insert to form a leak-free fluid path. Meets FDA requirements 21 CFR 177.1520(c) 3.2c and 21 CFR 176.170(c) and RoHS compliant
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Material Engage E8402
Packaging Quantity 10/Pk.
Inlet Female Luer, Port Disk
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.