Learn More
Cole-Parmer Cleanroom Port Disk Female Luer Fitting, Flat Plate, Engage™ 8402
Shop All Cole-Parmer Products
Click to view available options
:
10 Port Disk Luer Fitting - No Dimple Engage 8402
Description
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Fle in., ible, smooth flat base for easy attachment. Ultrasonic, radio frequency, or thermal weld to polyethylene bags. Connect to a male insert to form a leak-free fluid path. Meets FDA requirements 21 CFR 177.1520(c) 3.2c and 21 CFR 176.170(c) and RoHS compliant
Specifications
Specifications
| Certifications/Compliance | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Material | Engage E8402 |
| Packaging Quantity | 10/Pk. |
| Inlet | Female Luer, Port Disk |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.