Learn More
Nickel Electro™ Clifton™ Stainless Steel Raised Shelf
Obtain a good even heat distibution with perforted shelves. Nickel Electro™ Clifton™ Stainless Steel Raised Shelf allows two immersion depths by inverting the shelf.
Brand: Nickel Electro™ RS4-38
Additional Details : Weight : 1.00000kg
Description
- Simple solution for placing shallow vessels in a stirred bath
- Manufactured from stainless steel
- The raised shelf is perforated to allow free circulation of water
- Can be easily removed and cleaned
- Shelf can be inverted to provide two different immersion depths
Specifications
38 Liter Baths | |
Stainless Steel | |
60 x 35 x 20 cm |
Shelf Raised | |
60 x 35 x 20 cm |