Learn More
Nickel Electro™ Clifton™ Stainless Steel Raised Shelf
Obtain a good even heat distibution with perforted shelves. Nickel Electro™ Clifton™ Stainless Steel Raised Shelf allows two immersion depths by inverting the shelf.
Brand:  Nickel Electro™ RS4-38
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12768898
  • 312.00€ / Each
Estimated Shipment: 25-07-2024
to see stock.
Add to basket
Description
Description
  • Simple solution for placing shallow vessels in a stirred bath
  • Manufactured from stainless steel
  • The raised shelf is perforated to allow free circulation of water
  • Can be easily removed and cleaned
  • Shelf can be inverted to provide two different immersion depths
Specifications
Specifications
38 Liter Baths
Stainless Steel
60 x 35 x 20 cm
Shelf Raised
60 x 35 x 20 cm