Learn More
STEMCELL Technologies ClonaCell™-TCS/G418 Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
- This kit is used for selection and cloning applications where the G418 antibiotic is used as the selective agent
- ClonaCell™-TCS Medium is a serum-containing, methylcellulose-based semi-solid medium that can be used for selection and cloning of a variety of suspension-adapted and adherent cell lines, such as BHK-1, CHO-K1, CHO-S, CHO-DG44, and HEK-293
- This medium contains pre-selected fetal bovine serum (FBS) and bovine serum albumin (BSA)
- The G418 antibiotic is suitable for use in cell culture for the selection and maintenance of G418-resistant clones when using ClonaCell™-TCS Medium
Specifications
Specifications
| Cell Line | CHO Cells, HEK-293, Other |
| For Use With (Application) | Cell Culture; Semi-Solid Cloning |
| Quantity | 1 Kit |