missing translation for 'onlineSavingsMsg'
Learn More

Tocris™ CM 10

Product Code. p-7166831
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18749026

Brand: Tocris™ 6933/10

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

ALDH1A inhibitor; depletes CD133+ cancer stem cells (CSC)

CM 10 is a aldehyde dehydrogenase 1A (ALDH1A) inhibitor (IC50 values are 0.64, 0.74 and 1.7 μM for ALDH1A3, ALDH1A2 and ALDH1A1, respectively). Induces a necroptotic phenotype and preferentially depletes CD133+ ovarian cancer stem cells in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Molecular Formula C20H23N3O
Formula Weight 321.42
Product Type Inhibitor
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.