Learn More
Eppendorf™ Galaxy™ 48 R Incubators
When a high level of cell culture isolation is required or very limited space is available in the lab, the stackable Galaxy 48R is the right choice. The high temperature disinfection, a seamless chamber and a fanless design makes it easy to clean and provides high contamination protection.
Brand:  Eppendorf™ CO48310001
Additional Details : Weight : 30.00000kg
 View more versions of this product
Product Code. 15385194
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • 48 Liter (1.7 ft3) provides mid-sized capacity within a small footprint
  • Options: High Temp. Disinfection (4 hour at 120 °C), O2 control (e.g. for hypoxic studies), 2-split inner glass door
  • Fanless design offers more usable space
  • Six-sided direct heating system provides a uniform incubated environment
  • Deep-drawn, easy-to-clean stainless steel chamber without seams or welds
  • Low gas consumption
  • Integrated alarm relay
Specifications
Specifications
UK/HKG Power Supply
IR Sensor
None
Single Glass Inner Door
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products