Learn More
Buerkle™ Thread adapters LaboPlastBuerkle™ 3/4 in.
Shop All Bürkle Products
Click to view available options
Thread Size:
3/4 in. Captive Cap
3/4 in. Inflowing, 2 in. Mauser Outflowing
3/4 in. Inflowing, 2 in. Tri-Sure Outflowing
3/4 in. Inflowing, Conical R 3/4 in. Outflowing
Cylindrical G 3/4 in. External, 3/4 in. Internal
Fine Thread 2 in. External, 3/4 in. Internal
Hunersdorff External, 3/4 in. Internal
Kautex External, 3/4 in. Internal
Color:
Black
Blue
Gray
Green
Green/White
Red
Unit Size:
Each
Description
- All adapters have a standard outer thread R 3/4 in.
Specifications
Specifications
| Description | Thread Adapter 0.75 in., conical R 0.75 in. |
| Material | Polypropylene |
| Thread Size | 3/4 in. Inflowing, Conical R 3/4 in. Outflowing |
| Color | Green |
| Product Type | Thread Adapter |