missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) 2-methoxyethoxide, 99% (metals basis), 5% w/v in 2-methoxyethanol, Thermo Scientific Chemicals

Catalog Number p-7051449
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
142600-61-1
C6H14CoO4
209.107
MFCD00798554
BFBNUELICQBJBC-UHFFFAOYSA-N
cobalt ii 2-methoxyethoxide
131873636
cobalt(2+);2-methoxyethanolate
COCC[O-].COCC[O-].[Co+2]
This item is not returnable. View return policy
142600-61-1
C6H14CoO4
209.107
MFCD00798554
BFBNUELICQBJBC-UHFFFAOYSA-N
cobalt ii 2-methoxyethoxide
131873636
cobalt(2+);2-methoxyethanolate
COCC[O-].COCC[O-].[Co+2]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown
Density 0.96 g/mL
Boiling Point 124°C
Flash Point 40°C (104°F)
Linear Formula Co(OCH2CH2OCH3)2
Quantity 25 mL
UN Number UN1188
Sensitivity Moisture sensitive
Formula Weight 209.11
Percent Purity 99%
Concentration or Composition (by Analyte or Components) 5% w/v in 2-methoxyethanol
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Cobalt(II) 2-methoxyethoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'