Learn More
behr Labor-Technik™ COD Digestor System
Capable of simultaneous EPA, ISO, or DIN method digestion of up to 8/12 COD samples. behr Labor-Technik™ COD Digestor System is a workstation comprising all necessary components for COD sample digestion.
7020.00€ - 8225.00€
Specifications
Product Type | COD Digestor |
---|---|
Includes | Time/temperature regulator, heating block, holding frame, cooling bath, sample vessels, air-cooled condensers, condenser stand |
Product Code | Brand | No. of Places | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Places | Price | Quantity & Availability | |||||
10074130
|
behr Labor-Technik™
B00218405 |
8 |
7020.00€
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
10772001
|
behr Labor-Technik™
B00218406 |
12 |
8225.00€
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
Description
- Systems consist of the following components:
- Microprocessor-controlled TRS 200 time/temperature regulator with COD program
- Precision heating block CSB/E for RG2 vessels
- Holding frame E/B for RG2 sample vessels
- Cooling bath KW/N with mounting for E/B frame
- Sample vessels RG 2
- Air-cooled condensers LK 1
- Condenser stand LS for LK condensers
Specifications
COD Digestor | |
Time/temperature regulator, heating block, holding frame, cooling bath, sample vessels, air-cooled condensers, condenser stand |