Learn More
Masterflex™ Coiled FEP Tubing
Product Code. 11737278
Click to view available options
Diameter (Metric) Outer:
9.52 mm
Unit Size:
Each
Product Code. 11737278
Brand: Masterflex™ 9582103
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specifications
Autoclavable Autoclavable
Diameter (Metric) Inner 6.4 mm
Diameter (Metric) Outer 9.52 mm
Overall Length (Metric) 1.2 m
Min. Operating Temperature (Metric) -270°C
Packaging Quantity 1 Each
Sterility Sterile
Color Translucent
Maximum Pressure 318 psi
Max. Operating Temperature (Metric) 205°C