missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Native COL3A1. Source: Bovine Placenta (Accession #: P04258) The Collagen III alpha 1/COL3A1 Native Protein has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry.
Specifications
Specifications
| Accession Number | P04258 |
| For Use With (Application) | ELISA, Immunohistochemistry, SDS-PAGE, Western Blot |
| Formulation | 0.5 M Acetic Acid |
| Gene ID (Entrez) | 1281 |
| Name | Collagen III alpha 1/COL3A1 protein |
| Source | Bovine Serum |
| Storage Requirements | 4°C. Do not freeze. |
| Cross Reactivity | Bovine |
| Research Category | Cancer, Cell Biology, Extracellular Matrix |
| Purity or Quality Grade | Chromatography |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?