Learn More
Retsch™ Collecting Receptacle
Product Code. 15839757 Shop All Retsch Products
Click to view available options
For Use With (Equipment):
Cross Beater Mill SK 300
Cutting Mill SM 200
Unit Size:
Each
Product Code. 15839757
Brand: Retsch™ 22.003.0014
Only null left
Add to basket
Add to basket
Retsch™ Collecting Receptacle provides an easy way to collect the materials being ground up in cutting and cross beater mills.
  • Stainless steel
  • Holds up to 5 L
Specifications
Product Type Collecting Receptacle
Capacity (Metric) 5 L
For Use With (Equipment) Cutting Mill SM 200
Includes Adapter lid
Material Stainless Steel