missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Vivacon™ 500 are centrifugal units for fast DNA concentration and buffer exchange of 500μl samples. They are also the product of choice for protein fractionation applications. The use of Hydrosart™ membranes results in high recoveries from dilute samples. Complete recovery of the concentrate is guaranteed by a reverse spinning step.
Specifications
Specifications
| Color | Clear |
| Filter Type | Ultrafiltration |
| For Use With (Equipment) | Vivacon 500 |
| Sterility | Non-sterile |
| Includes | 100 reservoirs |
| Type | Tubing |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction