Learn More
Lovibond™ AF327 Platinum-Cobalt/Hazen/APHA Colour Test Kit
The Lovibond™ AF327 is a special kit for visual colour grading of Platinum-Cobalt/Hazen/APHA Colour over teh range 0 to 70mg Pt/l.
Brand: Lovibond™ 433270
Additional Details : Weight : 3.00000kg
Description
- Allows user to comply with the path length requirements of ASTM D 1209, which specifies a system for colour comparison where a 100mL column of sample 275 to 295mm in length is matched with platinum-cobalt solutions
- Typically used for light yellow liquids such as water, oils and chemicals
- The colour is determined by visually matching a column of sample in Nessler cylinder with precalibrated coloured glass standards, which are stable-coloured and not affected by UV light or extreme environmental conditions
- Because the sample colours are relatively unsaturated, the kit employs a long path length Nessleriser comparator to give greater measurement sensitivity and accuracy
Specifications
ASTM D 1209 | |
Applications: Beers, Malts and Caramels, Food and Beverages, Household Products, Medical and Clinical, Petroleum Oils and Waxes, Edible Oils and Fats, Sugar Solutions, Syrups and Honey, Pharmaceuticals and Cosmetics, Textiles, Coatings, Plastics, Chemicals, Swimming Pools and Spas, Water and Waste Waters | |
288 mm |
Color Measurement Test Kit | |
AF327 |