Learn More
VITLAB™ Colored LDPE Narrow Mouth Wash Bottles
Supply precise and small quantities of various liquids with these wash bottles. VITLAB™ Colored LDPE Narrow Mouth Wash Bottles feature a precise spray jet and optimized medium backflow through the continuous and finely drawn tip of the spray tube.
Brand:  VITLAB™ 132608
Additional Details : Weight : 0.13000kg
 View more versions of this product
Product Code. 11308613
  • 26.40€ / Pack of 5
Estimated Shipment: 03-06-2024
to see stock.
Add to basket
Description
Description
  • The VENT-CAP (patent pending) virtually prevents leaking, dripping or escaping of liquid from the dispensing tube, which can occur because of temperature changes in the laboratory
  • Narrow-mouth bottles, made from LDPE
  • Available in four different colors to facilitate ready identification
  • Screw cap and spray tube from PP
Specifications
Specifications
Wash Bottle
Vented
250 mL
LDPE
Narrow
135 mm
58 mm