missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Columbia Agar with Chocolated Horse Blood (Slope)

Product Code. 12726949
This item is not returnable. View return policy

Product Code. 12726949

Brand: Thermo Scientific™ BO0341B

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Isolate Haemophilus spp. from clinical specimens using these ready prepared slopes.

Isolate Haemophilus spp. from clinical specimens with Thermo Scientific™ Oxoid™ Columbia Agar with Chocolated Horse Blood Slopes. Chocolated horse blood is added to the general purpose Columbia Agar Base to supply the required nutrients for the growth of Haemophilus.

Remove the need for autoclaving and dispensing with convenient, pre-filled and ready prepared bottles of Columbia Agar with Chocolated Horse Blood Slopes.

  • Supports the growth of Haemophilus with chocolated blood providing the nutritional requirements.
  • Convenient: the pre-filled slopes are ready for immediate use.
  • Simple: easy-to-use with minimum training.
  • Long shelf life: up to 12 months.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Brown, opaque
Content And Storage 2°C to 10°C
Description Columbia Agar with Chocolated Horse Blood (Slope)
Format Bijou bottles
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 3.0g + 0.3g
Packaging Type Two hundred bijou bottles with screw cap closures in a box
Quantity 200/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.