Learn More
OHAUS™ Column Clamp
Product Code. 15702369 Shop All Ohaus Products
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
Each
Product Code. 15702369
Brand: OHAUS™ 30392316
Only null left
Add to basket
Add to basket
OHAUS™ Column Clamp is designed to securely grip and position a variety of laboratory glassware.
  • Corrosion-resistant stainless steel clamps provide excellent chemical resistance, durability, and can be autoclaved. Nickel-plated zinc clamps offer high-tensile strength at an economical price.
  • Prevent accidents in the lab! Versatile multi-purpose LabJaws clamps securely hold glassware and irregularly-shaped objects. Select from a variety of grip sizes, prong styles and adjustment types.
  • Choose from a variety of specialized LabJaws clamps with adjustable features to provide an exact fit for lab ware such as thermometers, thermocouples, columns, funnels, and round bottom flasks.
TRUSTED_SUSTAINABILITY
Specifications
Type Column Clamp
Material Stainless Steel
Description Clamp, Specialty, Column, CLS-COLMNSL
Diameter (Metric) 11 mm
Length (English) 13.31 in.
Length (English) Arm 8.310 in.
Length (Metric) 338.00 mm
Length (Metric) Arm 211 mm
Size Large
Size (Metric) Grip 64 to 140 mm
Jaw Type Column