Learn More
CombiCheck™ 10 Standard Solution Ammonium, Chloride, COD, Nitrate, Phosphate, Sulfate, WTW™
Click to view available options
Quantity:
100 mL
Unit Size:
Each
Description
- Matching concentration to test kit
- No calculation errors
- Stocking solution
Specifications
Specifications
| Solution Contains | Ammonium, Chloride, COD, Nitrate, Phosphate, Sulfate |
| Physical Form | Liquid |
| Concentration | Ammonium 4 mg/L, Chloride 25 mg/L, COD 80 mg/L, Nitrate 2.5 mg/L , Phosphate 0.8 mg/L, Sulfate 100 mg/L |
| Color | Colorless |
| pH | 5 |
| Boiling Point | 100°C |
| Solubility | Soluble in water |
| Quantity | 100 mL |