missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for HU6 Mini Horizontal Gel Electrophoresis Unit

For use with HU6 Mini Horizontal Gel Electrophoresis Unit

Brand:  Fisherbrand™

Additional Details : Weight : 2.00000kg

 View more versions of this product

Product Code. 10707131

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

  • Color coded
  • Height adjustable
  • Offer complete control over loading volume
  • Well depth to maximum 32-sample throughput
Specifications

Specifications

HU6 Mini Horizontal Gel Electrophoresis Unit
16
15 μL
Comb
1.5 mm
2.2 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions