missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for OmniPAGE Mini Vertical Gel Electrophoresis Unit, 0.75mm Thick

Product Code. 11867583 Shop All Fisherbrand Products
Click to view available options
No. of Wells:
1 Prep, 1 Marker
10
12
16
20
5
8
9
Volume (Metric) Well:
15 μL
20 μL
28 μL
30 μL
35 μL
40 μL
70 μL
500 μL
Unit Size:
Each

Product Code. 11867583

Only null left
Add to basket
Add to basket

For use in preparatory electrophoresis and high-throughput screening.

  • Accessory for use with OmniPAGE mini vertical gel electrophoresis unit
  • Recommended for ultra resolved bands
  • 8-sample and 16-sample combs are Multichannel pipettor compatible

Specifications

Color Black
For Use With (Application) Mini SDS PAGE, native PAGE, gradient, second dimension and nucleic acid separations
For Use With (Equipment) OmniPAGE Mini Vertical Gel Electrophoresis Unit
Type Comb
No. of Wells 10
Quantity 1 Each
Thickness (Metric) 0.75 mm
Volume (Metric) Well 30 μL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.