Learn More
Abeba™ Comfort 36872 Shoes
Stylish hand-sewn seams and PU sole with high slip resistance.
Brand:  Abeba™ 36872/40
Additional Details : Weight : 0.49000kg
 View more versions of this product
Product Code. 15670748
  • 97.46€ / 1 pair
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, E SRB
  • Velours
  • Black
  • Slip resistance PU outsole sole
  • Replaceable Reflexor insole with nubs (article no. 3572)
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Lining microfiber, respiration absorbing
  • Instep area with elastic band
  • Buckle fastening
  • Hand-sewn seam
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E SRB
Microfiber
40
Polyurethane
Black
Velour
Slip-resistant
36872