Learn More
Abeba™ Comfort 6819 Shoes
Stylish hand-sewn seams and PU sole with high slip resistance.
Brand: Abeba™ 6819/42
Additional Details : Weight : 0.85000kg
Description
- CE EN ISO 20347:2012 OB A, E SRB
- Smooth leather
- Black
- Slip resistance PU outsole sole
- Replaceable Reflexor insole with nubs (article no. 3572)
- Antistatic
- Lining microfiber, respiration absorbing
- 2 adjustable instep straps
- Double velcro fastening
- Hand-sewn seam
Specifications
CE EN ISO 20347:2012 OB A, E SRB | |
Microfiber | |
42 | |
Polyurethane |
Black | |
Leather | |
Slip-resistant | |
6819 |