Learn More
Abeba™ Comfort 6819 Shoes
Stylish hand-sewn seams and PU sole with high slip resistance.
Brand:  Abeba™ 6819/38
Additional Details : Weight : 0.85000kg
 View more versions of this product
Product Code. 15652338
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20347:2012 OB A, E SRB
  • Smooth leather
  • Black
  • Slip resistance PU outsole sole
  • Replaceable Reflexor insole with nubs (article no. 3572)
  • Antistatic
  • Lining microfiber, respiration absorbing
  • 2 adjustable instep straps
  • Double velcro fastening
  • Hand-sewn seam
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E SRB
Microfiber
38
Polyurethane
Black
Leather
Slip-resistant
6819