Learn More
Brand Compact automatic burette 25:0.05ml, BLAUBRAND, AS, DE-M, blue Schellbach stripe
Brand:  Brand 23924
Compact automatic burette 25:0.05ml, BLAUBRAND, AS, DE-M, blue Schellbach stripe
Product Code. 17265126
  • ONLINE EXCLUSIVE PRICE
    120.00€
    94.20€ / Each
    Save 25.80€
    21% Off
Only null left
Estimated Shipment: 14-01-2026