Learn More
Mecmesin™ Compact Force Gauge
The Compact Force Gauge (CFG+) is the entry-level, low-cost digital force gauge.
POA
Specifications
Height (Metric) 133 mm
Width (Metric) 76 mm
Weight (Metric) 250 g
View More Specs
Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
11938019
View Documents Promotion Details
Mecmesin™
860022
200 N
POA
Each
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
11974068
View Documents Promotion Details
Mecmesin™
860023
500 N
POA
Each
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
  • The CFG+ digital force gauge represents a highly affordable alternative to mechanical gauges and features an accuracy of ±0.5% of full-scale.
  • A clear simple LCD display shows readings in the most common measurement units (N, kN, mN, gf, kgf, lbf and ozf).
  • Values are shown as continuously changing ‘live’ or frozen in the display to capture the peak reading.
  • Manufactured in a lightweight ABS plastic housing, the CFG+ has a simple keypad and comes equipped with RS232 output for easy data transmission.
  • Its primary use is as a hand-held device but its design makes it attachable to manual test stands for more controlled testing.
  • An overload warning function, to reduce the risk of accidental damage to the instrument by an operator, is standard on all models.
Specifications
Specifications
133 mm
76 mm
250 g