Learn More
UVP Compact UV Lamp
Product Code. 11738241
Click to view available options
Product Type:
Compact UV Curing Lamp
Compact UV Lamp
Financial UV Lamp
Unit Size:
Each
Récupel-bijdrage uitgesloten
Product Code. 11738241
Brand: UVP 95001914
Only null left
Add to basket
Add to basket
Récupel-bijdrage uitgesloten
Specifications
Voltage 12 V
Wattage 4 W
Wavelength 365 nm
Dimensions (L x W) 198 x 71 mm
Length (Metric) 198 mm
Material Plastic
Model UVL-23R
Width (Metric) 71 mm
Product Type Compact UV Curing Lamp