Learn More
Thermo Scientific™ Complete Manual Staining Assembly 100
Product Code. 12667696 Shop All Epredia Products
Click to view available options
Product Type:
Staining Dish
Staining Dish Cover
Unit Size:
Each
Product Code. 12667696
Brand: Thermo Scientific™ 101
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Accommodate up to 19 slides with the Thermo Scientific™ Complete Staining Assembly 100, featuring a non-autoclavable glass dish.
Accommodate up to 19 slides with the Thermo Scientific™ Complete Staining Assembly 100, featuring a non-autoclavable glass dish.
Includes:
  • 109 rack/handle
  • 102 dish
  • 101 cover
Staining Rack 109:
Made from stainless steel and holds up to 19 slides. Maximum slide size is 3 x 2 in. (75 x 50mm).
Staining Dish 102:
Made from non-autoclavable glass with inside dimensions of 3.5 x 2.75 x 2.55 in. (90 x 70 x 65mm). Holds up to 250mL of solution with a fully loaded staining rack.
Staining Dish Cover 101:
Made from stainless steel.
TRUSTED_SUSTAINABILITY
Specifications
Includes Complete Assembly 100 includes 109 Rack/Handle, 102 Dish, and 101 Cover.
Product Type Staining Dish Cover