Learn More
Kern Sohn Microscope à lumière transmise OBN
Code produit 17369531
Encompass_Preferred
Modifier l'affichage
Click to view available options
Pouvoir grossissant:
10 fois, 20 fois, 40 fois, 100 fois
4 fois, 10 fois, 20 fois, 40 fois, 100 fois
Conditionnement:
1 pièce
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Pouvoir grossissant Conditionnement
17369531 10 fois, 20 fois, 40 fois, 100 fois 1 pièce
17329531 4 fois, 10 fois, 20 fois, 40 fois, 100 fois 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Règlementaire : Cet article est soumis à éco-contribution.
Code produit 17369531 Fournisseur Kern Sohn Code fournisseur OBN 158
 Article restreint
Estimated Shipment: 24-11-2026
Ajouter au panier
Ajouter au panier
missing translation for 'validMainframeMsg'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Règlementaire : Cet article est soumis à éco-contribution.
Professionalism and versatility united in one microscope - with Kohler illumination for demanding applications.
  • The OBN- series stands out because of its unbeatable and consistently high quality and its ergonomic design. The range of modular components means that the OBN series can be individually customised for the professional user.
  • Strong and continuously dimmable 3 W-LED illumination
  • This series has a professional Koehler illumination unit with an adjustable field diaphragm as well as a height-adjustable 1.25 Abbe condenser which can be centred and which has an adjustable aperture diaphragm.
  • The extremely large mechanical stage with ergonomic, coaxial coarse and fine focusing knob on both sides enables you to adjust and focus your sample rapidly and accurately
  • A wide variety of modular systems, such as, for example, a swing-out condenser, various eyepieces, objectives, colour filters, phase contrast units, a darkfield condenser, a simple polarising unit, a Butterfly Tube, through to complete fluorescence units are available to you as accessories
  • This centring eyepiece for adjusting the phase contrast, a protective dust cover, eye cups as well as multi-lingual user instructions are included in the scope of delivery
  • A C-mount adapter is required to connect a camera
  • Scope of application: haematology, urology, gynaecology, dermatology, pathology, microbiology and parasitology, immunology, Sewage treatment plants, oncology, entomology, Veterinary, water analysis, breweries
  • Applications/Samples: Translucent, thin, low-contrast, challenging samples (e.g. living mammal cells, bacteria, tissue)
Spécification
Type de tête Trinoculaire
Type d’éclairage Halogène
Pouvoir grossissant 10 fois, 20 fois, 40 fois, 100 fois
Tension 230 V